siRNA Details
| Gene Symbol: LOC101927314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110757.1 | 5 | 1636 | |||
| GenBank mRNA | AK091365.1 | - | 5 | 1585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927314| Gene Symbol: LOC101927314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110757.1 | 5 | 1636 | |||
| GenBank mRNA | AK091365.1 | - | 5 | 1585 | ||