siRNA Details
| Gene Symbol: FAM170B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038973.1 | 6 | 1534 | |||
| GenBank mRNA | AK093055.1 | - | 6 | 1536 | ||
| BC033360.1 | - | 3 | 608 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM170B-AS1| Gene Symbol: FAM170B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038973.1 | 6 | 1534 | |||
| GenBank mRNA | AK093055.1 | - | 6 | 1536 | ||
| BC033360.1 | - | 3 | 608 | |||