siRNA Details
| Gene Symbol: LOC101928475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135638.1 | 2 | 77 | |||
| GenBank mRNA | BC033345.1 | - | 2 | 77 | ||
| BG722758.1 | - | 2 | 82 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928475| Gene Symbol: LOC101928475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135638.1 | 2 | 77 | |||
| GenBank mRNA | BC033345.1 | - | 2 | 77 | ||
| BG722758.1 | - | 2 | 82 | |||