siRNA Details
| Gene Symbol: LOC102031319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104163.1 | 1, 2 | 118 | |||
| GenBank mRNA | BX107541.1 | - | 1, 2 | 118 | ||
| BX118505.1 | - | 1, 2 | 118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102031319| Gene Symbol: LOC102031319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104163.1 | 1, 2 | 118 | |||
| GenBank mRNA | BX107541.1 | - | 1, 2 | 118 | ||
| BX118505.1 | - | 1, 2 | 118 | |||