siRNA Details
| Gene Symbol: LOC101930010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125866.1 | 4 | 425 | |||
| GenBank mRNA | CK819990.1 | - | 4 | 425 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM273293.1 | - | 4 | 409 | Chr. 6 - 56844002 - 56864078 [+] on Build GRCh38 | |

LOC101930010
Genomic Map