siRNA Details
| Gene Symbol: LINC00632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028344.1 | 3 | 943 | |||
| GenBank mRNA | AK054978.1 | - | 3 | 935 | ||
| BQ343502.1 | - | 1 | 213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00632| Gene Symbol: LINC00632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028344.1 | 3 | 943 | |||
| GenBank mRNA | AK054978.1 | - | 3 | 935 | ||
| BQ343502.1 | - | 1 | 213 | |||