siRNA Details
Gene Symbol: LINC00632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028345.2 | 2 | 529 | |||
GenBank mRNA | BX508035.1 | - | 2 | 522 | ||
DA128399.1 | - | 2 | 529 | |||
DA529197.1 | - | 1 | 152 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028345.2 | 2 | 529 | |||
GenBank mRNA | BX508035.1 | - | 2 | 522 | ||
DA128399.1 | - | 2 | 529 | |||
DA529197.1 | - | 1 | 152 |