siRNA Details
| Gene Symbol: LOC105371766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135645.1 | 2 | 236 | |||
| GenBank mRNA | BX103390.1 | - | 2 | 236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371766| Gene Symbol: LOC105371766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135645.1 | 2 | 236 | |||
| GenBank mRNA | BX103390.1 | - | 2 | 236 | ||