siRNA Details
| Gene Symbol: LINC00850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109813.1 | 3 | 646 | |||
| GenBank mRNA | JX283354.1 | - | 3 | 646 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00850| Gene Symbol: LINC00850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109813.1 | 3 | 646 | |||
| GenBank mRNA | JX283354.1 | - | 3 | 646 | ||