siRNA Details
| Gene Symbol: LINC01270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034124.1 | 5 | 2042 | |||
| GenBank mRNA | AK090605.1 | - | 5 | 2042 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX100622.1 | - | 1 | 217 | Chr. 20 - 50314467 - 50314924 [+] on Build GRCh38 | |

LINC01270
Genomic Map