siRNA Details
| Gene Symbol: FAM155A-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046848.1 | 2 | 632 | |||
| GenBank mRNA | BC043519.1 | - | 2 | 624 | ||
| BI831413.1 | - | 2 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM155A-IT1| Gene Symbol: FAM155A-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046848.1 | 2 | 632 | |||
| GenBank mRNA | BC043519.1 | - | 2 | 624 | ||
| BI831413.1 | - | 2 | 629 | |||