siRNA Details
| Gene Symbol: CTB-113P19.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109873.1 | 5 | 1660 | |||
| GenBank mRNA | BC039364.1 | - | 5 | 1661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTB-113P19.1| Gene Symbol: CTB-113P19.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109873.1 | 5 | 1660 | |||
| GenBank mRNA | BC039364.1 | - | 5 | 1661 | ||