siRNA Details
| Gene Symbol: LOC101927211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110742.1 | 2, 3 | 232 | |||
| GenBank mRNA | BC032020.1 | - | 2, 3 | 232 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG719751.1 | - | 2, 3 | 236 | Chr. 6 - 57114901 - 57172213 [-] on Build GRCh38 | |

LOC101927211
Genomic Map