siRNA Details
| Gene Symbol: LOC101928424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135053.1 | 2 | 354 | |||
| GenBank mRNA | BX104024.1 | - | 2 | 340 | ||
| CR741883.1 | - | 2 | 355 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI792191.1 | - | 2 | 340 | Chr. 11 - 102452933 - 102461836 [+] on Build GRCh38 | |

LOC101928424
Genomic Map