siRNA Details
| Gene Symbol: PTOV1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110730.1 | 1 | 57 | |||
| GenBank mRNA | CK903988.1 | - | 1 | 57 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTOV1-AS2| Gene Symbol: PTOV1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110730.1 | 1 | 57 | |||
| GenBank mRNA | CK903988.1 | - | 1 | 57 | ||