siRNA Details
| Gene Symbol: LOC100128386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133571.1 | 2 | 592 | |||
| GenBank mRNA | AK124326.1 | - | 2 | 592 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI192871.1 | - | 2 | 296 | Chr. 11 - 100685180 - 100687659 [-] on Build GRCh38 | |

LOC100128386
Genomic Map