siRNA Details
| Gene Symbol: LOC100128386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133571.1 | 2 | 1709 | |||
| GenBank mRNA | AK124326.1 | - | 2 | 1709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100128386| Gene Symbol: LOC100128386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133571.1 | 2 | 1709 | |||
| GenBank mRNA | AK124326.1 | - | 2 | 1709 | ||