siRNA Details
| Gene Symbol: PCOLCE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038910.1 | 9 | 2522 | |||
| GenBank mRNA | AK097705.1 | - | 9 | 2522 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCOLCE-AS1| Gene Symbol: PCOLCE-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038910.1 | 9 | 2522 | |||
| GenBank mRNA | AK097705.1 | - | 9 | 2522 | ||