siRNA Details
| Gene Symbol: LOC102724502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135522.1 | 2 | 563 | |||
| GenBank mRNA | BX095520.1 | - | 2 | 563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724502| Gene Symbol: LOC102724502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135522.1 | 2 | 563 | |||
| GenBank mRNA | BX095520.1 | - | 2 | 563 | ||