siRNA Details
| Gene Symbol: CCDC144NL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104185.1 | 3 | 1616 | |||
| GenBank mRNA | AK057473.1 | - | 4 | 1876 | ||
| BC041488.2 | - | 3 | 1499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC144NL-AS1| Gene Symbol: CCDC144NL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104185.1 | 3 | 1616 | |||
| GenBank mRNA | AK057473.1 | - | 4 | 1876 | ||
| BC041488.2 | - | 3 | 1499 | |||