siRNA Details
| Gene Symbol: LOC101928557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135193.1 | 2 | 247 | |||
| GenBank mRNA | BX118346.1 | - | 2 | 247 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA383277.1 | - | 2 | 183 | Chr. 16 - 85792479 - 85792869 [+] on Build GRCh38 | |

LOC101928557
Genomic Map