siRNA Details
| Gene Symbol: LOC105377134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC024173.1 | - | 3 | 1102 | ||
| BC048201.1 | - | 3 | 831 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377134| Gene Symbol: LOC105377134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC024173.1 | - | 3 | 1102 | ||
| BC048201.1 | - | 3 | 831 | |||