siRNA Details
| Gene Symbol: LOC101927501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110387.1 | 6 | 1072 | |||
| GenBank mRNA | AK056105.1 | - | 6 | 1072 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927501| Gene Symbol: LOC101927501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110387.1 | 6 | 1072 | |||
| GenBank mRNA | AK056105.1 | - | 6 | 1072 | ||