siRNA Details
| Gene Symbol: LOC101928118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125948.1 | 1 | 154 | |||
| NR_125949.1 | 1 | 154 | ||||
| GenBank mRNA | AK090700.1 | - | 1 | 154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928118| Gene Symbol: LOC101928118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125948.1 | 1 | 154 | |||
| NR_125949.1 | 1 | 154 | ||||
| GenBank mRNA | AK090700.1 | - | 1 | 154 | ||