siRNA Details
| Gene Symbol: LINC01006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103858.1 | 1, 2 | 144 | |||
| GenBank mRNA | BM793926.1 | - | 1, 2 | 144 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01006| Gene Symbol: LINC01006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103858.1 | 1, 2 | 144 | |||
| GenBank mRNA | BM793926.1 | - | 1, 2 | 144 | ||