siRNA Details
| Gene Symbol: C7orf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026865.1 | 1 | 885 | |||
| GenBank mRNA | AF063598.1 | - | 1 | 652 | ||
| AF439977.1 | - | 1 | 885 | |||
| AK056371.1 | - | 1 | 801 | |||
| BC031037.1 | - | 1 | 770 | |||
| BC070228.1 | - | 2 | 210 | |||

C7orf13
Genomic Map