siRNA Details
| Gene Symbol: LINC00670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034144.1 | 3 | 912 | |||
| NR_034145.1 | 4 | 1074 | ||||
| GenBank mRNA | AK092009.1 | - | 4 | 1073 | ||
| BC122562.1 | - | 3 | 899 | |||
| BC122870.1 | - | 3 | 899 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00670| Gene Symbol: LINC00670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034144.1 | 3 | 912 | |||
| NR_034145.1 | 4 | 1074 | ||||
| GenBank mRNA | AK092009.1 | - | 4 | 1073 | ||
| BC122562.1 | - | 3 | 899 | |||
| BC122870.1 | - | 3 | 899 | |||