siRNA Details
| Gene Symbol: LOC102724659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126000.1 | 1 | 290 | |||
| GenBank mRNA | BC034298.1 | - | 1 | 290 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI826945.1 | - | 1 | 290 | Chr. 1 - 11778219 - 11779619 [-] on Build GRCh38 | |

LOC102724659
Genomic Map