siRNA Details
Gene Symbol: LOC105376633 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135110.1 | 3 | 277 | |||
GenBank mRNA | BX115137.1 | - | 3 | 278 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105376633 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135110.1 | 3 | 277 | |||
GenBank mRNA | BX115137.1 | - | 3 | 278 |