siRNA Details
| Gene Symbol: LOC101928012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110158.1 | 4 | 487 | |||
| GenBank mRNA | BC041883.1 | - | 4 | 490 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI561763.1 | - | 4 | 495 | Chr. 7 - 112622378 - 112699489 [+] on Build GRCh38 | |

LOC101928012
Genomic Map