siRNA Details
| Gene Symbol: LOC101927494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135563.1 | 2 | 577 | |||
| GenBank mRNA | BC035252.1 | - | 2 | 577 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI545414.1 | - | 2 | 582 | Chr. 3 - 86481943 - 86496439 [+] on Build GRCh38 | |

LOC101927494
Genomic Map