siRNA Details
| Gene Symbol: LOC101926913 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023515.1 | - | 3 | 755 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926913| Gene Symbol: LOC101926913 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023515.1 | - | 3 | 755 | ||