siRNA Details
Gene Symbol: LINC00662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027301.1 | 2 | 537 | |||
GenBank mRNA | AK055559.1 | - | 2 | 267 | ||
AK311452.1 | - | 2 | 537 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00662 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027301.1 | 2 | 537 | |||
GenBank mRNA | AK055559.1 | - | 2 | 267 | ||
AK311452.1 | - | 2 | 537 |