siRNA Details
| Gene Symbol: LOC100506302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126344.1 | 3 | 953 | |||
| GenBank mRNA | AK091942.1 | - | 1 | 2072 | ||
| DB517169.1 | - | 1 | 15 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506302| Gene Symbol: LOC100506302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126344.1 | 3 | 953 | |||
| GenBank mRNA | AK091942.1 | - | 1 | 2072 | ||
| DB517169.1 | - | 1 | 15 | |||