siRNA Details
| Gene Symbol: OVAAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125716.1 | 3 | 1415 | |||
| GenBank mRNA | BC036830.1 | - | 3 | 1412 | ||
| BC044260.1 | - | 3 | 1413 | |||
| BC063624.1 | - | 3 | 1415 | |||
| BC073892.1 | - | 3 | 1439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OVAAL| Gene Symbol: OVAAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125716.1 | 3 | 1415 | |||
| GenBank mRNA | BC036830.1 | - | 3 | 1412 | ||
| BC044260.1 | - | 3 | 1413 | |||
| BC063624.1 | - | 3 | 1415 | |||
| BC073892.1 | - | 3 | 1439 | |||