siRNA Details
Gene Symbol: OTUD6B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110438.1 | 3 | 813 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA812941.1 | - | 3 | 663 | Chr. 8 - 91063280 - 91070038 [-] on Build GRCh38 |
