siRNA Details
| Gene Symbol: LINC01490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120468.1 | 5 | 514 | |||
| NR_120469.1 | 5 | 458 | ||||
| NR_120470.1 | 3 | 310 | ||||
| GenBank mRNA | BC037383.1 | - | 3 | 314 | ||
| HY030171.1 | - | 5 | 424 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX109480.1 | - | 3 | 344 | Chr. 12 - 80764220 - 80769891 [+] on Build GRCh38 | |

LINC01490
Genomic Map