siRNA Details
| Gene Symbol: LOC101928782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110836.1 | 5 | 835 | |||
| GenBank mRNA | AK098603.1 | - | 5 | 834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928782| Gene Symbol: LOC101928782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110836.1 | 5 | 835 | |||
| GenBank mRNA | AK098603.1 | - | 5 | 834 | ||