siRNA Details
| Gene Symbol: LOC102723566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136302.1 | 6 | 2021 | |||
| GenBank mRNA | AK057719.1 | - | 6 | 2021 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723566| Gene Symbol: LOC102723566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136302.1 | 6 | 2021 | |||
| GenBank mRNA | AK057719.1 | - | 6 | 2021 | ||