siRNA Details
| Gene Symbol: LOC646268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033946.1 | 3 | 723 | |||
| GenBank mRNA | BC029599.1 | - | 3 | 722 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC646268| Gene Symbol: LOC646268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033946.1 | 3 | 723 | |||
| GenBank mRNA | BC029599.1 | - | 3 | 722 | ||