siRNA Details
| Gene Symbol: FLJ33581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040102.1 | 2 | 680 | |||
| GenBank mRNA | AK090900.1 | - | 2 | 680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ33581| Gene Symbol: FLJ33581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040102.1 | 2 | 680 | |||
| GenBank mRNA | AK090900.1 | - | 2 | 680 | ||