siRNA Details
Gene Symbol: LOC283683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040057.1 | 5 | 886 | |||
GenBank mRNA | BC044583.1 | - | 5 | 886 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC283683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040057.1 | 5 | 886 | |||
GenBank mRNA | BC044583.1 | - | 5 | 886 |