siRNA Details
Gene Symbol: ZNF529-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110703.1 | 2 | 499 | |||
NR_110704.1 | 2 | 529 | ||||
NR_110705.1 | 2 | 499 | ||||
NR_110706.1 | 2 | 499 | ||||
GenBank mRNA | BC039524.1 | - | 2 | 499 | ||
CD248632.1 | - | 2 | 177 | |||
CK820071.1 | - | 2 | 192 |
