siRNA Details
| Gene Symbol: BBOX1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125766.1 | 1, 2 | 437 | |||
| NR_125767.1 | 1, 2 | 77 | ||||
| NR_125768.1 | 1, 2 | 120 | ||||
| GenBank mRNA | DA376689.1 | - | 1, 2 | 89 | ||
| DA865512.1 | - | 1, 2 | 89 | |||
| DB016334.1 | - | 1, 2 | 77 | |||

BBOX1-AS1
Genomic Map