siRNA Details
| Gene Symbol: DACT3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040041.1 | 1 | 609 | |||
| GenBank mRNA | BC043434.2 | - | 1 | 609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DACT3-AS1| Gene Symbol: DACT3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040041.1 | 1 | 609 | |||
| GenBank mRNA | BC043434.2 | - | 1 | 609 | ||