siRNA Details
| Gene Symbol: LOC107133515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015341.2 | 1, 2 | 194 | |||
| GenBank mRNA | BG721507.1 | - | 1, 2 | 155 | ||
| DB024747.1 | - | 1, 2 | 194 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR740624.1 | - | 1, 2 | 160 | Chr. 17 - 30576499 - 30601284 [+] on Build GRCh38 | |

LOC107133515
Genomic Map