siRNA Details
| Gene Symbol: LOC101927843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109959.1 | 6 | 871 | |||
| GenBank mRNA | BC039377.1 | - | 6 | 871 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927843| Gene Symbol: LOC101927843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109959.1 | 6 | 871 | |||
| GenBank mRNA | BC039377.1 | - | 6 | 871 | ||