siRNA Details
| Gene Symbol: ARNTL2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109975.1 | 4 | 484 | |||
| GenBank mRNA | BU568793.1 | - | 4 | 487 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI832376.1 | - | 4 | 504 | Chr. 12 - 27389845 - 27446627 [-] on Build GRCh38 | |

ARNTL2-AS1
Genomic Map