siRNA Details
| Gene Symbol: TRIM52-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102762.1 | 3 | 613 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ187284.1 | - | 1 | 303 | Chr. 5 - 181271937 - 181272307 [+] on Build GRCh38 | |

TRIM52-AS1
Genomic Map