siRNA Details
| Gene Symbol: RBM26-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038991.1 | 2 | 213 | |||
| GenBank mRNA | AK002196.1 | - | 2 | 213 | ||
| BC034812.1 | - | 2 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBM26-AS1| Gene Symbol: RBM26-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038991.1 | 2 | 213 | |||
| GenBank mRNA | AK002196.1 | - | 2 | 213 | ||
| BC034812.1 | - | 2 | 174 | |||